Assay Details
Gene Symbol: LOC114116372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973017.1 | XP_027828818.1 | 2 - 3 | 471 | 103 | |
XM_027973018.1 | XP_027828819.1 | 2 - 3 | 474 | 103 |
Gene Symbol: LOC101103244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973014.1 | XP_027828815.1 | 27 - 28 | 3665 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
