Assay Details
Gene Symbol: LOC114116372 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027973017.1 | XP_027828818.1 | 8 - 9 | 1136 | 103 | |
XM_027973018.1 | XP_027828819.1 | 8 - 9 | 1139 | 103 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
