Assay Details
| Gene Symbol: NMRAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020746.4 | XP_004020795.2 | 2 - 3 | 169 | 55 | |
| XM_012104047.3 | XP_011959437.2 | 2 - 3 | 312 | 55 | ||
| XM_027962066.1 | XP_027817867.1 | 2 - 3 | 324 | 55 | ||
More Information
Set Membership: |
|

NMRAL1
Genomic Map