Assay Details
| Gene Symbol: LOC101112784 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004020955.4 | XP_004021004.1 | 3 - 4 | 416 | 114 | |
| XM_027962096.1 | XP_027817897.1 | 3 - 4 | 628 | 114 | ||
More Information
Set Membership: |
|

LOC101112784
Genomic Map