Assay Details
| Gene Symbol: HEY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004011142.4 | XP_004011191.2 | 2 - 3 | 190 | 83 | |
| GenBank mRNA | FJ712191.1 | - | 1 - 2 | 73 | 83 | |
More Information
Set Membership: |
|

HEY2
Genomic Map