Assay Details
| Gene Symbol: CBFA2T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977744.1 | XP_027833545.1 | 11 - 12 | 1810 | 61 | |
| XM_027977745.1 | XP_027833546.1 | 10 - 11 | 1733 | 61 | ||
| XM_027977746.1 | XP_027833547.1 | 11 - 12 | 1581 | 61 | ||
| XM_027977747.1 | XP_027833548.1 | 9 - 10 | 2273 | 61 | ||
| XM_027977748.1 | XP_027833549.1 | 11 - 12 | 1658 | 61 | ||
More Information
Set Membership: |
|

CBFA2T3
Genomic Map