Assay Details
| Gene Symbol: LOC101123244 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012101374.3 | XP_011956764.2 | 3 - 4 | 409 | 81 | |
| XM_027958714.1 | XP_027814515.1 | 3 - 4 | 403 | 81 | ||
| XM_027958715.1 | XP_027814516.1 | 3 - 4 | 401 | 81 | ||
| XM_027958716.1 | XP_027814517.1 | 3 - 4 | 407 | 81 | ||
More Information
Set Membership: |
|

LOC101123244
Genomic Map