Assay Details
| Gene Symbol: BICDL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027961754.1 | XP_027817555.1 | 7 - 8 | 1157 | 65 | |
| XM_027961755.1 | XP_027817556.1 | 7 - 8 | 1154 | 65 | ||
| XM_027961756.1 | XP_027817557.1 | 6 - 7 | 1314 | 65 | ||
More Information
Set Membership: |
|

BICDL2
Genomic Map