Assay Details
| Gene Symbol: FAM102B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002259.4 | XP_004002308.2 | 9 - 10 | 1023 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM102B| Gene Symbol: FAM102B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002259.4 | XP_004002308.2 | 9 - 10 | 1023 | 68 | |
Set Membership: |
|