Assay Details
| Gene Symbol: LOC101113317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006770.4 | XP_004006819.1 | 6 - 7 | 664 | 80 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101113317| Gene Symbol: LOC101113317 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006770.4 | XP_004006819.1 | 6 - 7 | 664 | 80 | |
Set Membership: |
|