Assay Details
| Gene Symbol: CHST10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012173773.2 | XP_012029163.1 | 6 - 7 | 637 | 62 | |
| XM_012173775.2 | XP_012029165.1 | 5 - 6 | 763 | 62 | ||
More Information
Set Membership: |
|

CHST10
Genomic Map