Assay Details
| Gene Symbol: LOC114112698 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027963847.1 | XP_027819648.1 | 2 - 3 | 152 | 96 | |
| Gene Symbol: EXOC6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012173630.2 | XP_012029020.2 | 4 - 5 | 541 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027963847.1 transcripts.
Set Membership: |
|

LOC114112698
Genomic Map