Assay Details
| Gene Symbol: DDX59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013934.4 | XP_004013983.2 | 6 - 7 | 1591 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DDX59| Gene Symbol: DDX59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004013934.4 | XP_004013983.2 | 6 - 7 | 1591 | 70 | |
Set Membership: |
|