Assay Details
| Gene Symbol: LOC114116650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973840.1 | XP_027829641.1 | 2 - 3 | 151 | 103 | |
| Gene Symbol: LOC101111560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973624.1 | XP_027829425.1 | 5 - 6 | 444 | 103 | |
More Information
Set Membership: |
|

LOC114116650
Genomic Map