Assay Details
| Gene Symbol: PRPS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962973.1 | XP_027818774.1 | 2 - 3 | 391 | 57 | |
| XM_027962974.1 | XP_027818775.1 | 2 - 3 | 391 | 57 | ||
More Information
Set Membership: |
|

PRPS2
Genomic Map