Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093333.2 | XP_014948819.2 | 9 - 10 | 1486 | 64 | |
| XM_027964789.1 | XP_027820590.1 | 10 - 11 | 1525 | 64 | ||
| XM_027964790.1 | XP_027820591.1 | 8 - 9 | 1881 | 64 | ||
| XM_027964791.1 | XP_027820592.1 | 8 - 9 | 1410 | 64 | ||
More Information
Set Membership: |
|

ZNF385B
Genomic Map