Assay Details
| Gene Symbol: LOC101120367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002175.4 | XP_004002224.1 | 5 - 6 | 854 | 155 | |
| XM_012175533.3 | XP_012030923.1 | 5 - 6 | 898 | 155 | ||
| XM_012175540.3 | XP_012030930.1 | 4 - 5 | 565 | 155 | ||
| XM_027971820.1 | XP_027827621.1 | 4 - 5 | 576 | 155 | ||
| XM_027971826.1 | XP_027827627.1 | 4 - 5 | 469 | 155 | ||
More Information
Set Membership: |
|

LOC101120367
Genomic Map