Assay Details
| Gene Symbol: LOC101120367 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002175.4 | XP_004002224.1 | 7 - 8 | 1381 | 79 | |
| XM_012175533.3 | XP_012030923.1 | 7 - 8 | 1425 | 79 | ||
| XM_012175540.3 | XP_012030930.1 | 6 - 7 | 1092 | 79 | ||
| XM_027971820.1 | XP_027827621.1 | 6 - 7 | 1103 | 79 | ||
| XM_027971826.1 | XP_027827627.1 | 6 - 7 | 996 | 79 | ||
More Information
Set Membership: |
|

LOC101120367
Genomic Map