Assay Details
| Gene Symbol: CREB3L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008602.4 | XP_004008651.1 | 9 - 10 | 1170 | 60 | |
| XM_012178073.3 | XP_012033463.1 | 9 - 10 | 1167 | 60 | ||
| XM_012178074.3 | XP_012033464.1 | 9 - 10 | 1126 | 60 | ||
| XM_012178075.3 | XP_012033465.1 | 8 - 9 | 1041 | 60 | ||
More Information
Set Membership: |
|

CREB3L3
Genomic Map