Assay Details
| Gene Symbol: RBKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004005779.3 | XP_004005828.1 | 6 - 7 | 709 | 58 | |
| XM_012166977.2 | XP_012022367.1 | 6 - 7 | 846 | 58 | ||
| XM_012166988.3 | XP_012022378.1 | 6 - 7 | 844 | 58 | ||
| XM_027966632.1 | XP_027822433.1 | 6 - 7 | 845 | 58 | ||
More Information
Set Membership: |
|

RBKS
Genomic Map