Assay Details
| Gene Symbol: BIRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015974.4 | XP_004016023.2 | 3 - 4 | 2383 | 62 | |
| XM_012095319.3 | XP_011950709.2 | 3 - 4 | 2383 | 62 | ||
More Information
Set Membership: |
|

BIRC2
Genomic Map