Assay Details
| Gene Symbol: DNAI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023217.4 | XP_004023266.2 | 6 - 7 | 955 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DNAI4| Gene Symbol: DNAI4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004023217.4 | XP_004023266.2 | 6 - 7 | 955 | 118 | |
Set Membership: |
|