Assay Details
| Gene Symbol: LOC105613870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012177743.3 | XP_012033133.1 | 3 - 4 | 393 | 86 | |
| XM_027979044.1 | XP_027834845.1 | 3 - 4 | 393 | 86 | ||
| XM_027979045.1 | XP_027834846.1 | 3 - 4 | 445 | 86 | ||
More Information
Set Membership: |
|

LOC105613870
Genomic Map