Assay Details
| Gene Symbol: RCN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015373.3 | XP_004015422.2 | 5 - 6 | 789 | 53 | |
| XM_027978619.1 | XP_027834420.1 | 5 - 6 | 703 | 53 | ||
More Information
Set Membership: |
|

RCN3
Genomic Map