Assay Details
| Gene Symbol: LOC101114383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015865.4 | XP_004015914.4 | 5 - 6 | 1044 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114383| Gene Symbol: LOC101114383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004015865.4 | XP_004015914.4 | 5 - 6 | 1044 | 68 | |
Set Membership: |
|