Assay Details
| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021753.4 | XP_004021802.1 | 2 - 3 | 194 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LEPROTL1| Gene Symbol: LEPROTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004021753.4 | XP_004021802.1 | 2 - 3 | 194 | 98 | |
Set Membership: |
|