Assay Details
| Gene Symbol: KCNMB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958237.1 | XP_027814038.1 | 2 - 3 | 294 | 71 | |
| GenBank mRNA | JF499671.1 | - | 1 - 2 | 149 | 71 | |
More Information
Set Membership: |
|

KCNMB2
Genomic Map