Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965088.1 | XP_027820889.1 | 11 - 12 | 1319 | 80 | |
| XM_027965089.1 | XP_027820890.1 | 11 - 12 | 1053 | 80 | ||
| XM_027965091.1 | XP_027820892.1 | 9 - 10 | 1091 | 80 | ||

FLACC1
Genomic Map