Assay Details
| Gene Symbol: FLACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965088.1 | XP_027820889.1 | 4 - 5 | 675 | 89 | |
| XM_027965089.1 | XP_027820890.1 | 4 - 5 | 409 | 89 | ||
| XM_027965090.1 | XP_027820891.1 | 4 - 5 | 675 | 89 | ||

FLACC1
Genomic Map