Assay Details
| Gene Symbol: TPRKB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006077.3 | XP_004006126.1 | 2 - 3 | 884 | 98 | |
| XM_004006078.4 | XP_004006127.1 | 2 - 3 | 321 | 98 | ||
| XM_012173645.2 | XP_012029035.1 | 2 - 3 | 863 | 98 | ||
| XM_012173646.2 | XP_012029036.1 | 2 - 3 | 263 | 98 | ||
| XM_012173647.3 | XP_012029037.1 | 2 - 3 | 318 | 98 | ||
More Information
Set Membership: |
|

TPRKB
Genomic Map