Assay Details
| Gene Symbol: CXCL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015098600.2 | XP_014954086.1 | 3 - 4 | 880 | 85 | |
| XM_027975073.1 | XP_027830874.1 | 4 - 5 | 688 | 85 | ||
More Information
Set Membership: |
|

CXCL16
Genomic Map