Assay Details
| Gene Symbol: MBIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017899.4 | XP_004017948.1 | 2 - 3 | 322 | 76 | |
| XM_012098826.3 | XP_011954216.1 | 2 - 3 | 322 | 76 | ||
| XM_015102012.2 | XP_014957498.1 | 2 - 3 | 333 | 76 | ||
More Information
Set Membership: |
|

MBIP
Genomic Map