Assay Details
| Gene Symbol: POLR1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018844.4 | XP_004018893.1 | 6 - 7 | 737 | 67 | |
| XM_012100703.3 | XP_011956093.1 | 6 - 7 | 737 | 67 | ||
More Information
Set Membership: |
|

POLR1C
Genomic Map