Assay Details
| Gene Symbol: DYNC2I1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969086.1 | XP_027824887.1 | 21 - 22 | 2578 | 98 | |
| XM_027969087.1 | XP_027824888.1 | 24 - 25 | 3035 | 98 | ||
| XM_027969088.1 | XP_027824889.1 | 21 - 22 | 2837 | 98 | ||
More Information
Set Membership: |
|

DYNC2I1
Genomic Map