Assay Details
| Gene Symbol: DENND2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979401.1 | XP_027835202.1 | 18 - 19 | 4016 | 85 | |
| XM_027979402.1 | XP_027835203.1 | 19 - 20 | 3520 | 85 | ||
| XM_027979403.1 | XP_027835204.1 | 18 - 19 | 6348 | 85 | ||
| XM_027979404.1 | XP_027835205.1 | 18 - 19 | 3615 | 85 | ||

DENND2B
Genomic Map