Assay Details
| Gene Symbol: SYT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015100319.2 | XP_014955805.2 | 3 - 4 | 945 | 52 | |
| XM_027978798.1 | XP_027834599.1 | 2 - 3 | 582 | 52 | ||
| XM_027978799.1 | XP_027834600.1 | 3 - 4 | 476 | 52 | ||
More Information
Set Membership: |
|

SYT5
Genomic Map