Assay Details
| Gene Symbol: LOC554335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974354.1 | XP_027830155.1 | 21 - 22 | 3295 | 101 | |
| XM_027974355.1 | XP_027830156.1 | 15 - 16 | 2261 | 101 | ||
| XM_027974356.1 | XP_027830157.1 | 10 - 11 | 1905 | 101 | ||
| GenBank mRNA | AJ920032.1 | - | 10 - 11 | 1537 | 101 | |
More Information
Set Membership: |
|

LOC554335
Genomic Map