Assay Details
| Gene Symbol: LOC114113217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027965152.1 | XP_027820953.1 | 1 - 2 | 39 | 87 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_027965152.1 transcripts.
Set Membership: |
|

LOC114113217
Genomic Map