Assay Details
| Gene Symbol: LOC101114011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008130.4 | XP_004008179.1 | 3 - 4 | 356 | 85 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101114011| Gene Symbol: LOC101114011 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008130.4 | XP_004008179.1 | 3 - 4 | 356 | 85 | |