Assay Details
| Gene Symbol: LOC101120576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971377.1 | XP_027827178.1 | 9 - 10 | 445 | 89 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101120576| Gene Symbol: LOC101120576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027971377.1 | XP_027827178.1 | 9 - 10 | 445 | 89 | |