Assay Details
| Gene Symbol: LOC114115243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970755.1 | XP_027826556.1 | 5 - 6 | 575 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC114115243| Gene Symbol: LOC114115243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970755.1 | XP_027826556.1 | 5 - 6 | 575 | 99 | |
Set Membership: |
|