Assay Details
| Gene Symbol: SAP30BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974081.1 | XP_027829882.1 | 5 - 6 | 449 | 62 | |
| XM_027974083.1 | XP_027829884.1 | 5 - 6 | 487 | 62 | ||
| XM_027974084.1 | XP_027829885.1 | 5 - 6 | 448 | 62 | ||
| XM_027974085.1 | XP_027829886.1 | 4 - 5 | 401 | 62 | ||
More Information
Set Membership: |
|

SAP30BP
Genomic Map