Assay Details
| Gene Symbol: SLC3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019620.4 | XP_004019669.2 | 6 - 7 | 1186 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC3A2| Gene Symbol: SLC3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019620.4 | XP_004019669.2 | 6 - 7 | 1186 | 69 | |
Set Membership: |
|