Assay Details
| Gene Symbol: ACP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004002403.4 | XP_004002452.2 | 9 - 10 | 1602 | 62 | |
| XM_027973906.1 | XP_027829707.1 | 10 - 11 | 1799 | 62 | ||
More Information
Set Membership: |
|

ACP6
Genomic Map