Assay Details
| Gene Symbol: MACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004007779.4 | XP_004007828.1 | 2 - 3 | 272 | 66 | |
| XM_012176466.3 | XP_012031856.1 | 3 - 4 | 373 | 66 | ||
| XM_027968502.1 | XP_027824303.1 | 2 - 3 | 997 | 66 | ||
More Information
Set Membership: |
|

MACC1
Genomic Map