Assay Details
| Gene Symbol: SLC1A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979148.1 | XP_027834949.1 | 8 - 9 | 1454 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC1A6| Gene Symbol: SLC1A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979148.1 | XP_027834949.1 | 8 - 9 | 1454 | 58 | |
Set Membership: |
|