Assay Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009558.3 | XP_004009607.1 | 3 - 4 | 309 | 69 | |
| XM_012178978.3 | XP_012034368.1 | 3 - 4 | 460 | 69 | ||
More Information
Set Membership: |
|

RFESD
Genomic Map