Assay Details
| Gene Symbol: COASY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012931.4 | XP_004012980.2 | 7 - 8 | 1515 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

COASY| Gene Symbol: COASY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004012931.4 | XP_004012980.2 | 7 - 8 | 1515 | 62 | |
Set Membership: |
|