Assay Details
| Gene Symbol: SYDE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015095779.2 | XP_014951265.2 | 5 - 6 | 1452 | 62 | |
| XM_027979144.1 | XP_027834945.1 | 6 - 7 | 1326 | 62 | ||
More Information
Set Membership: |
|

SYDE1
Genomic Map