Assay Details
| Gene Symbol: MAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009048.4 | XP_004009097.1 | 6 - 7 | 963 | 76 | |
| XM_004009049.4 | XP_004009098.1 | 6 - 7 | 957 | 76 | ||
More Information
Set Membership: |
|

MAT2B
Genomic Map